Cyclopropanesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR12688-100G
OR12688-25G
OR12688-5G
APOSOR12688-100GEA
3540
CZK
APOSOR12688-100G
APOSOR12688-25G
APOSOR12688-5G
Cyclopropanesulphonyl chloride
Cyclopropanesulphonyl chloride
Vzorec:
C₃H₅ClO₂S Bod varu: 60 °C/20mmHg Hustota: 1,39 g/cm³ (25 °C) Bod vzplanutí: 96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01631933 CAS číslo: 139631-62-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...