(3,5-Dichlorophenyl)methanesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,5-Dichloro-α-toluenesulphonyl chloride
OR12682-1G
OR12682-250MG
APOSOR12682-1GEA
2280
CZK
APOSOR12682-1G
APOSOR12682-250MG
(3,5-Dichlorophenyl)methanesulphonyl chloride
(3,5-Dichlorophenyl)methanesulphonyl chloride
Vzorec:
C₇H₅CI₃O₂S MW: 259,54 g/mol Bod tání: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631928 CAS číslo: 163295-70-3 UN: 3096 ADR: 8+4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...