Tisk…

4'-Methylbiphenyl-4-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4'-Methyl-[1,1'-biphenyl]-4-sulphonyl chloride

OR12669-5G OR12669-1G
APOSOR12669-5GEA 5590 CZK
APOSOR12669-5G APOSOR12669-1G
4'-Methylbiphenyl-4-sulphonyl chloride
4'-Methylbiphenyl-4-sulphonyl chloride
Vzorec: C₁₃H₁₁ClO₂S
MW: 266,74 g/mol
Bod tání: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631922
CAS číslo: 101366-51-2
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR