4-Phenoxybenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Phenoxybenzene-1-sulphonyl chloride
OR12614-100MG
OR12614-250MG
APOSOR12614-100MGEA
622
CZK
APOSOR12614-100MG
APOSOR12614-250MG
4-Phenoxybenzenesulphonyl chloride
4-Phenoxybenzenesulphonyl chloride
Vzorec:
C₁₂H₉ClO₃S MW: 268,72 g/mol Bod varu: 180 °C/5mmHg Bod tání: 41…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00625748 CAS číslo: 1623-92-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...