Tisk…

3-Phenoxybenzylamine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Phenoxyphenyl)methylamine hydrochloride

OR12556-1G OR12556-5G
APOSOR12556-1GEA 4930 CZK
APOSOR12556-1G APOSOR12556-5G
3-Phenoxybenzylamine hydrochloride 97%
3-Phenoxybenzylamine hydrochloride
Vzorec: C₁₃H₁₄ClNO
MW: 235.72 g/mol
Bod tání: 185.5…187 °C
Storage Temperature: Ambient
MDL Number: MFCD07781045
CAS číslo: 376637-85-3
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR