Tisk…

5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR1255-1G OR1255-100MG
APOSOR1255-1GEA 3150 CZK
APOSOR1255-1G APOSOR1255-100MG
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulphonyl chloride
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulphonyl chloride
Vzorec: C₈H₆ClNO₂S₃
MW: 279,79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00828611
CAS číslo: 215434-25-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR