Tisk…

2-(2-Pyrrolidinyl)pyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Pyrrolidin-2-ylpyridine

OR12515-1G OR12515-250MG
APOSOR12515-1GEA 4370 CZK
APOSOR12515-1G APOSOR12515-250MG
2-(2-Pyrrolidinyl)pyridine 95%
2-(2-Pyrrolidinyl)pyridine
Vzorec: C₉H₁₂N₂
MW: 148.21 g/mol
Bod varu: 82 °C/0.8mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD01691538
CAS číslo: 77790-61-5
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR