Tisk…

1-[α-(4,6-Dimethoxypyrimidin-2-yl)benzyl]piperazine 95+%

Dodavatel: Apollo Scientific

OR1248-10G OR1248-1G OR1248-5G
APOSOR1248-10GEA 45630 CZK
APOSOR1248-10G APOSOR1248-1G APOSOR1248-5G
1-[α-(4,6-Dimethoxypyrimidin-2-yl)benzyl]piperazine 95+%
1-[alpha-(4,6-Dimethoxypyrimidin-2-yl)benzyl]piperazine
Vzorec: C₁₇H₂₂N₄O₂
MW: 314,39 g/mol
Bod tání: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD01765673
CAS číslo: 354563-89-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR