Tisk…

6-Chloroimidazo[2,1-b][1,3]thiazole

Dodavatel: Apollo Scientific
Varování

OR1246-1G OR1246-5G
APOSOR1246-1GEA 2070 CZK
APOSOR1246-1G APOSOR1246-5G
6-Chloroimidazo[2,1-b][1,3]thiazole
6-Chloroimidazo[2,1-b][1,3]thiazole
Vzorec: C₅H₃ClN₂S
MW: 158,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173802
CAS číslo: 23576-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR