1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Methyl-1H-benzo[d]imidazole-5-carboxylic acid
OR12418-5G
APOSOR12418-5GEA
4370
CZK
APOSOR12418-5G
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid 97%
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid
Vzorec:
C₉H₈N₂O₂ MW: 176,18 g/mol Bod tání: 293…295 °C Storage Temperature: Ambient |
MDL Number:
MFCD06809665 CAS číslo: 53484-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...