1-Methyl-1H-pyrazole-5-carbaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR12399-1G
OR12399-25G
OR12399-5G
APOSOR12399-1GEA
622
CZK
APOSOR12399-1G
APOSOR12399-25G
APOSOR12399-5G
1-Methyl-1H-pyrazole-5-carbaldehyde
1-Methyl-1H-pyrazole-5-carbaldehyde
Vzorec:
C₅H₆N₂O Bod varu: 115 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03419801 CAS číslo: 27258-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...