Tisk…

Methyl-3-(chlorocarbonyl)benzoate 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR12388-1G OR12388-5G
APOSOR12388-1GEA 4570 CZK
APOSOR12388-1G APOSOR12388-5G
Methyl-3-(chlorocarbonyl)benzoate 97%
Methyl-3-(chlorocarbonyl)benzoate
Vzorec: C₉H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD09702420
CAS číslo: 3441-03-0
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR