Tisk…

tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, N-Boc-Nortropinone

OR12337-25G OR12337-100G OR12337-5G
APOSOR12337-25GEA 1080 CZK
APOSOR12337-25G APOSOR12337-100G APOSOR12337-5G
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 98%
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Vzorec: C₁₂H₁₉NO₃
MW: 225,29 g/mol
Bod tání: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00673779
CAS číslo: 185099-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR