Tisk…

(1-Methyl-1H-benzimidazol-2-yl)methylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Aminomethyl)-1-methyl-1H-benzimidazole, (1-Methyl-1H-benzo[d]imidazol-2-yl)methanamine

OR12313-1G OR12313-250MG OR12313-25G
APOSOR12313-1GEA 3380 CZK
APOSOR12313-1G APOSOR12313-250MG APOSOR12313-25G
(1-Methyl-1H-benzimidazol-2-yl)methylamine
(1-Methyl-1H-benzimidazol-2-yl)methylamine
Vzorec: C₉H₁₁N₃
MW: 161.21 g/mol
Bod tání: 261…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00457746
CAS číslo: 20028-40-4
UN: 3259
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR