Tisk…

Benzo[b]thiophene-7-carbonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR12299-1G OR12299-250MG
APOSOR12299-1GEA 6290 CZK
APOSOR12299-1G APOSOR12299-250MG
Benzo[b]thiophene-7-carbonyl chloride 97%
Benzo[b]thiophene-7-carbonyl chloride
Vzorec: C₉H₅ClOS
Bod tání: 84…86 °C
Storage Temperature: Ambient
CAS číslo: 120081-47-2
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR