Tisk…

5-Methyl-2-thiophenecarbonitrile 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Methylthiophene-2-carbonitrile, 2-Cyano-5-methylthiophene

OR12283-5G
APOSOR12283-5GEA 2030 CZK
APOSOR12283-5G
5-Methyl-2-thiophenecarbonitrile 95%
5-Methyl-2-thiophenecarbonitrile
Vzorec: C₆H₅NS
MW: 123,1756 g/mol
Bod varu: 73…75 °C/5mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD01434334
CAS číslo: 72835-25-7
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR