2-Chloro-3,5-dimethylpyrazine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR12278-1G
OR12278-5G
APOSOR12278-1GEA
622
CZK
APOSOR12278-1G
APOSOR12278-5G
2-Chloro-3,5-dimethylpyrazine 97%
2-Chloro-3,5-dimethylpyrazine
Vzorec:
C₆H₇ClN₂ MW: 142,5900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00126945 CAS číslo: 38557-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...