4-(2-Methylimidazol-1-yl)phenylamine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(2-Methyl-1H-imidazol-1-yl)aniline
OR12260-1G
OR12260-5G
OR12260-10G
APOSOR12260-1GEA
1210
CZK
APOSOR12260-1G
APOSOR12260-5G
APOSOR12260-10G
4-(2-Methylimidazol-1-yl)phenylamine 97%
4-(2-Methylimidazol-1-yl)phenylamine
Vzorec:
C₁₀H₁₁N₃ MW: 173,2144 g/mol Bod tání: 111…113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06797788 CAS číslo: 74852-81-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...