1-Methyl-2-oxo-2,3-dihydro-1H-indole-5-sulphonyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Methyl-2-oxoindoline-5-sulphonyl chloride, 1-Methyl-2-oxindole-5-sulphonyl chloride
OR12235-1G
OR12235-250MG
APOSOR12235-1GEA
4960
CZK
APOSOR12235-1G
APOSOR12235-250MG
1-Methyl-2-oxo-2,3-dihydro-1H-indole-5-sulphonyl chloride 95%
1-Methyl-2-oxo-2,3-dihydro-1H-indole-5-sulphonyl chloride
Vzorec:
C₉H₈ClNO₃S MW: 245,69 g/mol Bod tání: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD08691362 CAS číslo: 166883-20-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...