N-(1H-Indazol-7-yl)thiourea
Dodavatel: Apollo Scientific
OR12197-1G
OR12197-500MG
APOSOR12197-1GEA
15310
CZK
APOSOR12197-1G
APOSOR12197-500MG
N-(1H-Indazol-7-yl)thiourea
N-(1H-Indazol-7-yl)thiourea
Vzorec:
C₈H₈N₄S MW: 192,24 g/mol Bod tání: 203…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD03102719 CAS číslo: 381211-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...