N-[2-(Hydroxymethyl)phenyl]-4-iodobenzenesulphonamide
Dodavatel: Apollo Scientific
OR12196-1G
OR12196-5G
APOSOR12196-1GEA
7670
CZK
APOSOR12196-1G
APOSOR12196-5G
N-[2-(Hydroxymethyl)phenyl]-4-iodobenzenesulphonamide
N-[2-(Hydroxymethyl)phenyl]-4-iodobenzenesulphonamide
Vzorec:
C₁₃H₁₂INO₃S Storage Temperature: Ambient |
MDL Number:
MFCD09027185 CAS číslo: 952182-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...