Tisk…

4-Methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Methyl-2-(methylthio)pyrimidine-5-carboxylic acid

OR12144-1G OR12144-500MG OR12144-5G
APOSOR12144-1GEA 2360 CZK
APOSOR12144-1G APOSOR12144-500MG APOSOR12144-5G
4-Methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylic acid 95%
4-Methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylic acid
Vzorec: C₇H₈N₂O₂S
MW: 184,22 g/mol
Bod tání: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD02187391
CAS číslo: 98276-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR