Tisk…

Ethyl-2-(1-benzyl-4-piperidinylidene)acetate

Dodavatel: Apollo Scientific
Varování

OR12094-1G
APOSOR12094-1GEA 2900 CZK
APOSOR12094-1G
Ethyl-2-(1-benzyl-4-piperidinylidene)acetate
Ethyl-2-(1-benzyl-4-piperidinylidene)acetate
Vzorec: C₁₆H₂₁NO₂
MW: 259,35 g/mol
Bod varu: 120…123 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD08437068
CAS číslo: 40110-55-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR