3-(4-Bromo-3-methylphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Dodavatel: Apollo Scientific
OR12045-1G
APOSOR12045-1GEA
5130
CZK
APOSOR12045-1G
3-(4-Bromo-3-methylphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
3-(4-Bromo-3-methylphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Vzorec:
C₁₅H₁₁BrN₂OS MW: 347,24 g/mol Bod tání: 312…314 °C Storage Temperature: Ambient |
MDL Number:
MFCD00175729 CAS číslo: 883042-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...