Tisk…

3,5-Dimethyl-4-propoxyphenylboronic acid

Dodavatel: Apollo Scientific

Synonyms: (3,5-Dimethyl-4-propoxyphenyl)boronic acid

OR1203-100G OR1203-25G OR1203-5G
APOSOR1203-100GEA 15310 CZK
APOSOR1203-100G APOSOR1203-25G APOSOR1203-5G
3,5-Dimethyl-4-propoxyphenylboronic acid
3,5-Dimethyl-4-propoxyphenylboronic acid
Vzorec: C₁₁H₁₇BO₃
MW: 208,0619 g/mol
Bod tání: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD05865178
CAS číslo: 357611-51-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR