Tisk…

4-Bromo-1-isopropylpyrazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-1-isopropyl-1H-pyrazole

OR11985-5G
APOSOR11985-5GEA 1700 CZK
APOSOR11985-5G
4-Bromo-1-isopropylpyrazole
4-Bromo-1-isopropylpyrazole
Vzorec: C₆H₉BrN₂
MW: 189,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801021
CAS číslo: 313735-62-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR