2-Methyl-5-pyridineboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(6-Methylpyridin-3-yl)boronic acid
OR11980-1G
OR11980-25G
OR11980-5G
APOSOR11980-1GEA
744
CZK
APOSOR11980-1G
APOSOR11980-25G
APOSOR11980-5G
2-Methyl-5-pyridineboronic acid
2-Methyl-5-pyridineboronic acid
Vzorec:
C₆H₈BNO₂ MW: 136.94 g/mol Bod tání: 90…100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06801736 CAS číslo: 659742-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...