4-(6-Chloropyridazin-3-yl)morpholine
Dodavatel: Apollo Scientific
|
Varování
|
OR11964-1G
OR11964-5G
APOSOR11964-1GEA
612
CZK
APOSOR11964-1G
APOSOR11964-5G
4-(6-Chloropyridazin-3-yl)morpholine
4-(6-Chloropyridazin-3-yl)morpholine
Vzorec:
C₈H₁₀ClN₃O Storage Temperature: Ambient |
MDL Number:
MFCD00574575 CAS číslo: 17259-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...