Tisk…

2-(1H-Pyrazol-1-yl)ethanol

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(1-Pyrazolyl)ethanol

OR11946-1G OR11946-250MG
APOSOR11946-1GEA 1210 CZK
APOSOR11946-1G APOSOR11946-250MG
2-(1H-Pyrazol-1-yl)ethanol
2-(1H-Pyrazol-1-yl)ethanol
Vzorec: C₅H₈N₂O
MW: 112,13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00046076
CAS číslo: 6314-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR