7-Methylimidazo[1,2-a]pyridine
Dodavatel: Apollo Scientific
OR11945-1G
OR11945-5G
APOSOR11945-1GEA
2940
CZK
APOSOR11945-1G
APOSOR11945-5G
7-Methylimidazo[1,2-a]pyridine
7-Methylimidazo[1,2-a]pyridine
Vzorec:
C₈H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD06761695 CAS číslo: 874-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...