4-(2-Thiazolylcarbamoyl)phenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR1192-1G
OR1192-50MG
OR1192-5G
APOSOR1192-1GEA
6850
CZK
APOSOR1192-1G
APOSOR1192-50MG
APOSOR1192-5G
4-(2-Thiazolylcarbamoyl)phenylboronic acid 98%
4-(2-Thiazolylcarbamoyl)phenylboronic acid
Vzorec:
C₁₀H₉BN₂O₃S Bod tání: 206…210 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659877 CAS číslo: 850568-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...