3-Bromo-N,N-diisopropylbenzamide
Dodavatel: Apollo Scientific
OR11910-1G
OR11910-5G
OR11910-10G
APOSOR11910-1GEA
0
CZK
APOSOR11910-1G
APOSOR11910-5G
APOSOR11910-10G
3-Bromo-N,N-diisopropylbenzamide
3-Bromo-N,N-diisopropylbenzamide
Vzorec:
C₁₃H₁₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD00587757 CAS číslo: 35309-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...