4-Boronobenzenesulphonamide 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Sulphamoylbenzeneboronic acid, (4-Aminosulphonylphenyl)boronic acid
OR1186-5G
OR1186-1G
OR1186-10G
APOSOR1186-5GEA
8270
CZK
APOSOR1186-5G
APOSOR1186-1G
APOSOR1186-10G
4-Boronobenzenesulphonamide 95%
4-Boronobenzenesulphonamide
Vzorec:
C₆H₈BNO₄S MW: 201.00 g/mol Bod tání: 250…256 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659839 CAS číslo: 613660-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...