Tisk…

1,3-Dimethylpyrrolidin-2-one

Dodavatel: Apollo Scientific
Varování

OR11733-250MG OR11733-100MG OR11733-1G
APOSOR11733-250MGEA 3310 CZK
APOSOR11733-250MG APOSOR11733-100MG APOSOR11733-1G
1,3-Dimethylpyrrolidin-2-one
1,3-Dimethylpyrrolidin-2-one
Vzorec: C₆H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD01687329
CAS číslo: 19597-07-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR