Tisk…

5-(3-Nitrophenyl)-4-oxazolecarboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(3-Nitrophenyl)-1,3-oxazole-4-carboxylic acid

OR11681-10MG OR11681-50MG
APOSOR11681-10MGEA 622 CZK
APOSOR11681-10MG APOSOR11681-50MG
5-(3-Nitrophenyl)-4-oxazolecarboxylic acid
5-(3-Nitrophenyl)-4-oxazolecarboxylic acid
Vzorec: C₁₀H₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD09800992
CAS číslo: 951885-28-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR