Tisk…

1-(2-(4-Nitrophenoxy)ethyl)pyrrolidine

Dodavatel: Apollo Scientific

OR11670-1G OR11670-5G OR11670-25G
APOSOR11670-1GEA 3020 CZK
APOSOR11670-1G APOSOR11670-5G APOSOR11670-25G
1-(2-(4-Nitrophenoxy)ethyl)pyrrolidine
1-(2-(4-Nitrophenoxy)ethyl)pyrrolidine
Vzorec: C₁₂H₁₆N₂O₃
MW: 236.2700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01308390
CAS číslo: 265654-77-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR