1-(4-Bromobenzoyl)pyrrolidine
Dodavatel: Apollo Scientific
Synonyms:
(4-Bromophenyl)(pyrrolidin-1-yl)methanone
OR11626-5G
OR11626-25G
APOSOR11626-5GEA
2780
CZK
APOSOR11626-5G
APOSOR11626-25G
1-(4-Bromobenzoyl)pyrrolidine
1-(4-Bromobenzoyl)pyrrolidine
Vzorec:
C₁₁H₁₂BrNO MW: 254,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00595283 CAS číslo: 5543-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...