Tisk…

4-Bromo-N,N-diisopropylbenzamide

Dodavatel: Apollo Scientific

OR11619-1G OR11619-5G
APOSOR11619-1GEA 2610 CZK
APOSOR11619-1G APOSOR11619-5G
4-Bromo-N,N-diisopropylbenzamide
4-Bromo-N,N-diisopropylbenzamide
Vzorec: C₁₃H₁₈BrNO
MW: 284.1921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00591594
CAS číslo: 79606-46-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR