5-Bromo-2-chloronicotinonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-Bromo-2-chloro-3-cyanopyridine
OR11594-1G
OR11594-5G
APOSOR11594-1GEA
622
CZK
APOSOR11594-1G
APOSOR11594-5G
5-Bromo-2-chloronicotinonitrile
5-Bromo-2-chloronicotinonitrile
Vzorec:
C₆H₂BrClN₂ MW: 217.45 g/mol Bod tání: 135…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD09801046 CAS číslo: 405224-23-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...