Ethyl-8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR11589-1G
OR11589-5G
APOSOR11589-1GEA
1820
CZK
APOSOR11589-1G
APOSOR11589-5G
Ethyl-8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Vzorec:
C₁₀H₈BrClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09607953 CAS číslo: 951884-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...