5-Bromo-4-isooctylpyrimidine
Dodavatel: Apollo Scientific
Synonyms:
5-Bromo-4-(6-methylheptyl)pyrimidine
OR11583-1G
OR11583-5G
APOSOR11583-1GEA
3150
CZK
APOSOR11583-1G
APOSOR11583-5G
5-Bromo-4-isooctylpyrimidine
5-Bromo-4-isooctylpyrimidine
Vzorec:
C₁₂H₁₉BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09800941 CAS číslo: 951884-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...