Tisk…

4-Bromo-N-methylbenzamide 98%

Dodavatel: Apollo Scientific
Varování

OR11557-1G OR11557-25G OR11557-5G
APOSOR11557-1GEA 622 CZK
APOSOR11557-1G APOSOR11557-25G APOSOR11557-5G
4-Bromo-N-methylbenzamide 98%
4-Bromo-N-methylbenzamide
Vzorec: C₈H₈BrNO
MW: 214,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462943
CAS číslo: 27466-83-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR