Tisk…

6-Chloro-N-propyl-4-pyrimidinamine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Chloro-6-(propylamino)pyrimidine

OR11551-1G OR11551-10G OR11551-5G
APOSOR11551-1GEA 2900 CZK
APOSOR11551-1G APOSOR11551-10G APOSOR11551-5G
6-Chloro-N-propyl-4-pyrimidinamine 97%
6-Chloro-N-propyl-4-pyrimidinamine
Vzorec: C₇H₁₀ClN₃
MW: 171,63 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09475893
CAS číslo: 941294-33-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR