Tisk…

3-Bromo-N-cyclopropyl-5-nitrobenzamide 97%

Dodavatel: Apollo Scientific

OR11493-1G OR11493-5G
APOSOR11493-1GEA 3310 CZK
APOSOR11493-1G APOSOR11493-5G
3-Bromo-N-cyclopropyl-5-nitrobenzamide 97%
3-Bromo-N-cyclopropyl-5-nitrobenzamide
Vzorec: C₁₀H₉BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09475856
CAS číslo: 941294-17-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR