3,5-Dicarboxyphenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,5-Dicarboxybenzeneboronic acid, 5-Boronoisophthalic acid
OR11481-25G
OR11481-5G
OR11481-100G
APOSOR11481-25GEA
4960
CZK
APOSOR11481-25G
APOSOR11481-5G
APOSOR11481-100G
3,5-Dicarboxyphenylboronic acid 98%
3,5-Dicarboxyphenylboronic acid
Vzorec:
C₈H₇BO₆ MW: 209,95 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06203347 CAS číslo: 881302-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...