1-(p-Toluenesulphonyl)pyrrole 96%
Dodavatel: Apollo Scientific
Synonyms:
1-Tosylpyrrole, 1-Tosyl-1H-pyrrole, N-Tosylpyrrole
OR11477-100G
OR11477-25G
APOSOR11477-100GEA
6090
CZK
APOSOR11477-100G
APOSOR11477-25G
1-(p-Toluenesulphonyl)pyrrole 96%
1-(p-Toluenesulphonyl)pyrrole
Vzorec:
C₁₁H₁₁NO₂S MW: 221,27 g/mol Bod tání: 99…104,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145014 CAS číslo: 17639-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...