Tisk…

1-(2-Bromoethoxy)-4-nitrobenzene 95%

Dodavatel: Apollo Scientific

OR11447-25G OR11447-100G
APOSOR11447-25GEA 3060 CZK
APOSOR11447-25G APOSOR11447-100G
1-(2-Bromoethoxy)-4-nitrobenzene 95%
1-(2-Bromoethoxy)-4-nitrobenzene
Vzorec: C₈H₈BrNO₃
MW: 246,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031363
CAS číslo: 13288-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR