Tisk…

3-Bromo-5-methylaniline hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

OR11430-1G OR11430-5G
APOSOR11430-1GEA 622 CZK
APOSOR11430-1G APOSOR11430-5G
3-Bromo-5-methylaniline hydrochloride 98%
3-Bromo-5-methylaniline hydrochloride
Vzorec: C₇H₉BrClN
Storage Temperature: Ambient
MDL Number: MFCD09475832
CAS číslo: 957034-79-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR