Tisk…

4-Chloro-6-(1-pyrrolidinyl)pyrimidine 98%

Dodavatel: Apollo Scientific
Varování

OR11426-1G OR11426-5G OR11426-10G
APOSOR11426-1GEA 2820 CZK
APOSOR11426-1G APOSOR11426-5G APOSOR11426-10G
4-Chloro-6-(1-pyrrolidinyl)pyrimidine 98%
4-Chloro-6-(1-pyrrolidinyl)pyrimidine
Vzorec: C₈H₁₀ClN₃
Storage Temperature: Ambient
MDL Number: MFCD09475835
CAS číslo: 939986-64-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR