3,5-Dichloropyridine-2-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
OR11385-25G
OR11385-100G
OR11385-500G
APOSOR11385-25GEA
1120
CZK
APOSOR11385-25G
APOSOR11385-100G
APOSOR11385-500G
3,5-Dichloropyridine-2-carbonitrile
3,5-Dichloropyridine-2-carbonitrile
Vzorec:
C₆H₂Cl₂N₂ MW: 173,00 g/mol Bod tání: 98…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788758 CAS číslo: 85331-33-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...